Learn More
Cayman Chemical BB-22 6-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2704733-61-7, 10 mg
SDP
Catalog No. 502971072
Encompass_Preferred
Differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its six position; intended for forensic and research applications
Catalog No. 50-297-1072 Supplier Cayman Chemical Supplier No. 1454310 MG
Differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its six position; intended for forensic and research applications