Learn More
Cayman Chemical BB-22 7-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365471-55-0, 10 mg
SDP
Catalog No. 502970422
Encompass_Preferred
BB-22 7-hydroxyisoquinoline isomer differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its seven position; intended for forensic and research applications
Catalog No. 50-297-0422 Supplier Cayman Chemical Supplier No. 1454410 MG
BB-22 7-hydroxyisoquinoline isomer differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its seven position; intended for forensic and research applications