Learn More
Medchemexpress LLC BB-Cl-Amidine | 1802637-39-3 | 98.2% | 459.97 | 10 MG
SDP
Catalog No. 5000346336
Encompass_Preferred
BB-Cl-Amidine is a peptidylarginine deminase (PAD) inhibitor. It exhibits cytotoxicity in U2OS cells and has shown to improve endothelium-dependent vasorelaxation in MRL/lpr mice, subtly reducing splenomegaly and increasing anti-NET antibodies.
  • Acts as a peptidylarginine deminase (PAD) inhibitor
  • Exhibits cytotoxicity in U2OS cells with EC50/IC50 of 8.8 μM
  • Significantly improves endothelium-dependent vasorelaxation in animal models
  • Shows a strong trend towards downregulation of IRGs
  • Subtly reduces splenomegaly and increases anti-NET antibodies
  • Improves muzzle alopecia
  • Soluble in various solvents including DMSO, SBE-β-CD in Saline, Corn Oil, and PEG300/Tween-80/Saline
Catalog No. 50-003-46336 Supplier Medchemexpress LLC Supplier No. HY11134710MM/1ML
BB-Cl-Amidine is a peptidylarginine deminase (PAD) inhibitor. It exhibits cytotoxicity in U2OS cells and has shown to improve endothelium-dependent vasorelaxation in MRL/lpr mice, subtly reducing splenomegaly and increasing anti-NET antibodies.
  • Acts as a peptidylarginine deminase (PAD) inhibitor
  • Exhibits cytotoxicity in U2OS cells with EC50/IC50 of 8.8 μM
  • Significantly improves endothelium-dependent vasorelaxation in animal models
  • Shows a strong trend towards downregulation of IRGs
  • Subtly reduces splenomegaly and increases anti-NET antibodies
  • Improves muzzle alopecia
  • Soluble in various solvents including DMSO, SBE-β-CD in Saline, Corn Oil, and PEG300/Tween-80/Saline