Learn More
Medchemexpress LLC Bb-cl-amidine | 1802637-39-3 | MFCD31746879 | 98.2% | 459.97 g·mol⁻¹ | C26H26ClN5O | 5 MG
SDP
Catalog No. 5000263289
Encompass_Preferred
BB-Cl-Amidine is a small-molecule irreversible inhibitor of peptidylarginine deiminases (PADs) used in biochemical and cellular research to study protein arginine deimination. It is supplied as a solid research reagent with supporting analytical data and common salt options for varied solubility needs.
  • Irreversible PAD inhibitor suitable for biochemical and cellular assays.
  • High reported purity (≈98.2%).
  • Solid, white to off-white form for straightforward handling and storage.
  • Available in common salt forms (hydrochloride and TFA) to aid solubility choices.
  • Analytical data available, including COA, H NMR, HPLC, and MS.
  • For research use only; not for human or animal therapeutic use.
Catalog No. 50-002-63289 Supplier Medchemexpress LLC Supplier No. HY1113475MG
BB-Cl-Amidine is a small-molecule irreversible inhibitor of peptidylarginine deiminases (PADs) used in biochemical and cellular research to study protein arginine deimination. It is supplied as a solid research reagent with supporting analytical data and common salt options for varied solubility needs.
  • Irreversible PAD inhibitor suitable for biochemical and cellular assays.
  • High reported purity (≈98.2%).
  • Solid, white to off-white form for straightforward handling and storage.
  • Available in common salt forms (hydrochloride and TFA) to aid solubility choices.
  • Analytical data available, including COA, H NMR, HPLC, and MS.
  • For research use only; not for human or animal therapeutic use.