Learn More
Medchemexpress LLC HY-111347A 10mg Medchemexpress, BB-Cl-Amidine (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501968837
Encompass_Preferred
Medchemexpress, HY-111347A 10mg BB-Cl-Amidine (HCl) CAS: BB-Cl-Amidine hydrochloride is a peptidylarginine deminase (PAD) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8837 Supplier Medchemexpress LLC Supplier No. HY111347A10MG
Medchemexpress, HY-111347A 10mg BB-Cl-Amidine (HCl) CAS: BB-Cl-Amidine hydrochloride is a peptidylarginine deminase (PAD) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.