Learn More
Medchemexpress LLC BB-Cl-Amidine hydrochloride | 2436747-41-8 | 99.5% | 496.43 | 1 ML
SDP
Catalog No. 5000343148
Encompass_Preferred
BB-Cl-Amidine hydrochloride is a peptidylarginine deiminase (PAD) inhibitor intended for research use only. This compound has demonstrated significant biological activity, including improving endothelium-dependent vasorelaxation and reducing splenomegaly in MRL/lpr mice.
  • Subtly reduces splenomegaly in MRL/lpr mice
  • Shows a trend towards increased circulating levels of anti-NET antibodies
  • Improves endothelium-dependent vasorelaxation
  • Downregulates IRGs
  • Significantly improves muzzle alopecia
  • Administered at 1 mg/kg daily via subcutaneous injection in animal models
Catalog No. 50-003-43148 Supplier Medchemexpress LLC Supplier No. HY11134750MG
BB-Cl-Amidine hydrochloride is a peptidylarginine deiminase (PAD) inhibitor intended for research use only. This compound has demonstrated significant biological activity, including improving endothelium-dependent vasorelaxation and reducing splenomegaly in MRL/lpr mice.
  • Subtly reduces splenomegaly in MRL/lpr mice
  • Shows a trend towards increased circulating levels of anti-NET antibodies
  • Improves endothelium-dependent vasorelaxation
  • Downregulates IRGs
  • Significantly improves muzzle alopecia
  • Administered at 1 mg/kg daily via subcutaneous injection in animal models