Learn More
Thermco Products Inc Accu-Safe Enclosed Chamber Bottle thermometer, Blood bank/Refrigerator, Spirit, -5 deg./20 deg.C, 0.5 deg.C div, 145mm, 250ml bio-safe glycol/H2O
Catalog No. NC0626280
Encompass_Preferred
We’re currently building out the product information and will be adding additional content soon. Thank you for your patience as we continue to expand our selection and improve your ordering experience.
Catalog No. NC0626280 Supplier Thermco Products Inc Supplier No. ACCBBS