Learn More
Rapid Micro Biosystems Inc BB VISION LIDS 48/CASE
SDP
Catalog No. NC2582454
Encompass
  • Transparent, non-fluorescing plastic material
  • Designed for high-throughput image acquisition
  • Facilitates accurate detection and enumeration of microorganisms
  • Support non-destructive testing, preserving samples for further identification
  • Manufactured in an ISO9001 certified facility
  • Treated with ethylene oxide for sterilization
  • Case quantity of 48 lids
  • Storage at room temperature
  • Exclusive compatibility with Growth Direct system
  • Not intended for In Vitro Diagnostic use
Catalog No. NC2582454 Supplier Rapid Micro Biosystems Inc Supplier No. BBVL048
  • Transparent, non-fluorescing plastic material
  • Designed for high-throughput image acquisition
  • Facilitates accurate detection and enumeration of microorganisms
  • Support non-destructive testing, preserving samples for further identification
  • Manufactured in an ISO9001 certified facility
  • Treated with ethylene oxide for sterilization
  • Case quantity of 48 lids
  • Storage at room temperature
  • Exclusive compatibility with Growth Direct system
  • Not intended for In Vitro Diagnostic use