Learn More
Eurofins Discoverx ChemiScreen BB1 Bombesine/Neuromedin B Receptor Stable Cell Line, 2 vials. (HTS123C)
Catalog No. 502005926
Encompass
The ChemiScreen BB1 Bombesin/Neuromedin B Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bombesin, neuromedin B) based activation of NMBR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NMBR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NMBR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5926 Supplier Eurofins Discoverx Supplier No. HTS123C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen BB1 Bombesin/Neuromedin B Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bombesin, neuromedin B) based activation of NMBR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NMBR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NMBR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.