Learn More
Medchemexpress LLC BB2-50F | 2226086-65-1 | 99.5% | 393.44 | 5 MG
SDP
Catalog No. 5000392958
Encompass_Preferred
BB2-50F is an antimicrobial agent and succinate dehydrogenase and F1Fo-ATP synthase inhibitor.
  • Induces ROS.
  • Inhibits succinate oxidation and reduces tricarboxylic acid (TCA) cycle activity.
  • Inhibits the growth of Mycobacterium tuberculosis H37Rv and its auxotrophic, attenuated derivative mc2 6230 (ΔRD1, ΔpanCD) with an MIC of 8 μM.
Catalog No. 50-003-92958 Supplier Medchemexpress LLC Supplier No. HY1621425MG
BB2-50F is an antimicrobial agent and succinate dehydrogenase and F1Fo-ATP synthase inhibitor.
  • Induces ROS.
  • Inhibits succinate oxidation and reduces tricarboxylic acid (TCA) cycle activity.
  • Inhibits the growth of Mycobacterium tuberculosis H37Rv and its auxotrophic, attenuated derivative mc2 6230 (ΔRD1, ΔpanCD) with an MIC of 8 μM.