Learn More
Cayman Chemical BB-22 3-carboxyindole metabolite, Purity: ≥98%, CAS: 858515-71-6, 10 mg
SDP
Catalog No. 502972131
Encompass_Preferred
An expected metabolite of BB-22 that would be detectable both in serum and urine; intended for forensic and research applications
Catalog No. 50-297-2131 Supplier Cayman Chemical Supplier No. 1437810 MG
An expected metabolite of BB-22 that would be detectable both in serum and urine; intended for forensic and research applications