Learn More
Cayman Chemical BB22 4hydroxyIsoquInolIn I 1mg
SDP
Catalog No. 502970665
Encompass_Preferred
Differs from BB-22 structurally by having an isoquinoline ring attached at the four position instead of a quinoline attached through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-297-0665 Supplier Cayman Chemical Supplier No. 145411 MG
Differs from BB-22 structurally by having an isoquinoline ring attached at the four position instead of a quinoline attached through the eight position of the ring; intended for forensic and research applications