Learn More
Cayman Chemical BB22 7hydroxyIsoquInolIn I 1mg
SDP
Catalog No. 502971525
Encompass_Preferred
BB-22 7-hydroxyisoquinoline isomer differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its seven position; intended for forensic and research applications
Catalog No. 50-297-1525 Supplier Cayman Chemical Supplier No. 145441 MG
BB-22 7-hydroxyisoquinoline isomer differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its seven position; intended for forensic and research applications