Learn More
Eurofins Discoverx ChemiScreen BB3 Bombesin Receptor Membrane Preparation, 1 vial. (HTS160M)
Catalog No. 502005989
Encompass
The ChemiScreen BB3 Bombesin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of BRS3 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5989 Supplier Eurofins Discoverx Supplier No. HTS160M
The ChemiScreen BB3 Bombesin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of BRS3 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.