Learn More
Description
Host Species:
Species Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | Streptavidin |
| Conjugate | BB515 |
| Cross Reactivity | Mouse |
| Quantity | 0.1 mg |
| Form | Affinity-purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.