Learn More
BD Cell Analysis 3P BB700 Mouse Anti-Human CD25, 100 tests
Supplier:  BD Cell Analysis 3P 566447
Encompass
Vol. Per Test: 5 µl, Isotype: Mouse BALB/c IgG1, κ; Reactivity: Human (QC Testing) Rhesus, Cynomolgus, Baboon (Tested in Development), Application: Flow cytometry (Routinely Tested), Immunogen: Phytohemagglutinin stimulated human lymphocytes, Alternative Name: IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55
Catalog No. NC1649925