Learn More
Aviva Systems Biology Corp BBC3 Antibody - N-terminal region (ARP58945_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58945_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BBC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BBC3. Alternate names: FLJ42994, JFY1, PUMA, JFY-1ApplicationWB. Species reactivity: Human
Catalog No. 50-286-536