Learn More
Lonza Walkersville Bovine Brain Extract (BBE), 5 ml
Catalog No. NC9500151
Encompass
Bovine Brain Extract is useful in the culture of various endothelial cell types. The product is offered at a concentration of 9 mg/mL. Content:1 x Bottle containing 9 mg/mL Bovine Brain Extract, 5 mL
Catalog No. NC9500151 Supplier Lonza Walkersville Supplier No. CC4098
Bovine Brain Extract is useful in the culture of various endothelial cell types. The product is offered at a concentration of 9 mg/mL. Content:1 x Bottle containing 9 mg/mL Bovine Brain Extract, 5 mL