Learn More
Lonza Walkersville Bovine Brain Extract (BBE), 2 ml
Catalog No. NC2077160
Encompass
Bovine Brain Extract is offered at a concentration of 3 mg/ml and is sold individually or as a component in Lonza's SingleQuots Kits associated with endothelial cell media.Content:1 x Bottle containing 3 mg/mL BBE, 2 mL
Catalog No. NC2077160 Supplier Lonza Walkersville Supplier No. CC4092
Bovine Brain Extract is offered at a concentration of 3 mg/ml and is sold individually or as a component in Lonza's SingleQuots Kits associated with endothelial cell media.Content:1 x Bottle containing 3 mg/mL BBE, 2 mL