Learn More
BD BBL™ Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K1 Prepared Media
Shop All BD Diagnostic Systems ProductsSpecifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Red |
| Additive | Hemin, Vitamin K1 |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Agar with 5% Sheep Blood, Hemin and Vitamin K1 |
| Product Type | Prepared Microbiology Plate |
| Formulation | Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, Sheep Blood, defibrinated 5% |
| For Use With (Application) | Fastidious, obligately anaerobic organisms |
| Packaging Type | Plate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.