Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Crystal™ ID Systems Accessories

Catalog No. B4345029 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Microbiology Media
Quantity:
10 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
B4345029 Microbiology Media 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B4345029 Supplier BD Supplier No. 245029

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BBL Crystal MIND Software

  • Interprets the test results and provides specimen identification
  • Minimum software requirements are an MS-DOS™ 5.0 or higher compatible computer; Intel™ 486 microprocessor with at least 16MB RAM Software is compatible with Microsoft™ Windows™ (3.11, 95, 98, 2000, XP) Computer must have at least Microsoft ODBC Version 2.00 and a 3.5 in. floppy drive
Codebooks Include Enteric/Nonfermenter, Anaerobe, Neisseria/Haemophilus, Gram-Positive, Rapid Gram-Positive) Supplied in individual hard-copy binder BBL Crystal Panel Viewer
  • For facilitated viewing and recording of chromogenic and fluorogenic reactions
BBL CrystalSpec™ Standards†
  • Set of two calibration standards for the CrystalSpec Nephelometer, one Calibration Blank and one Calibration Standard

†Drop shipment is required. All shipments FOB from Cockeysville, MD. Freight charges will be prepaid and billed as a separate item on the invoice.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Microbiology Media
For Use With (Equipment) Crystal™ ID system
Quantity 10 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.