Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Phosphate Buffered Saline

Catalog No. DF2314150 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
Packaging Type:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF2314-15-0 100 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF2314-15-0 Supplier BD Supplier No. 223142
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorescent Antibody reagents

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Additive Phosphate Buffered Saline
Form Dehydrated
For Use With (Application) Fluorescent Antibody reagents
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.