Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Fluid Thioglycollate Medium Sterile Pack Bottles (ETO)

Catalog No. B257097 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Fluid thioglycollate medium; Sterile pack bottle; Quantity: 100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B257097 Fluid thioglycollate medium; Sterile pack bottle; Quantity: 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B257097 Supplier BD Supplier No. 257097
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for sterility testing of biologics and for enrichment and cultivation of anaerobes, aerobes and microaerophiles

  • The entire package unit is sealed in two Stericlin™ bags and sterilized by ethylene oxide (ETO) treatment.
  • This process ensures a sterile outer surface of the containers and package.
  • The multiple bags allow for the introduction of the bottles into cleanrooms from a non-sterile to a sterile area.
  • Used for the sterility testing of biologics.
  • Used for the enrichment and cultivation of anaerobes, aerobes, and microaerophiles.
  • Specially developed clear FTM for much easier growth detection.
  • The headspace is gassed with a stream of nitrogen to maintain anaerobiosis.
TRUSTED_SUSTAINABILITY

Specifications

Appearance Light Yellow to Amber, Clear to Slightly Hazy, Without Precipitate or Debris
Broth Type Fluid Thioglycollate Medium
Cap Type Crimp
Certifications/Compliance USP
Color Light Yellow to Amber
Additive Sodium Thioglycollate
Detects Microorganisms, Anaerobes
Form Liquid
Format Bottle
Media Type Fluid
Product Type Liquid Medium
Sterility Sterile
Target Organism Class Bacteria
Test Type Sterility Testing
Fill Volume 100 mL
Final Product Type Microorganisms
Formulation Yeast Extract-5.0 g, Casitone Peptone-15.0 g, Dextrose-5.5 g, NaCl-2.5 g, L-Cystine-0.5 g, Sodium thioglycollate-0.5 g, Agar-0.75 g, Resazurin-1.0 mg, DI Water-1000 ml
For Use With (Application) Sterility Testing, Cultivation of Anaerobes, Aerobes, and Microaerophiles
Includes Infusion Bottle, Crimp Cap with Tear Tab
Packaging Type Bottle
Product Line BBL
Quantity 100 mL
Specimen Type Biologics, Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.