Learn More
BD BBL™ Gram Stain Kits
GSA_VA
Catalog No. B12539 Shop All Waters Corporation Products
Change view
Click to view available options
Product Type:
Gram Stain Kit (Stabilized)
Gram Stain Kit (Unstabilized)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B12539 Gram Stain Kit (Stabilized)
B12524 Gram Stain Kit (Unstabilized)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12539 Supplier BD Supplier No. 212539
 Restricted Item
Add to Cart
Add to Cart
Used to stain microorganisms from cultures or specimens by the differential Gram method
  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Each break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Certifications/Compliance CE
Includes 250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Stain Type Gram Stain
Volume (Metric) 250 mL
Product Type Gram Stain Kit (Stabilized)