Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media

Catalog No. BB21716 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 x 2 mL
100 x 5 mL
100 x 8 mL
100 x 10 mL
100 x 15 mL
Packaging Type:
Size A tubes with cap
Size K tubes with cap
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BB21716 100 x 5 mL Size K tubes with cap
BB21093 100 x 8 mL Size K tubes with cap
BB21815 100 x 2 mL Size K tubes with cap
B97354X 100 x 10 mL Size K tubes with cap
B21823X 100 x 15 mL Size A tubes with cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. BB21716 Supplier BD Supplier No. 221716
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP
Color Beige
Content And Storage 15°C to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase Soy Broth
Product Type Prepared Microbiology Tubes
Sterility Sterile
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
For Use With (Application) For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms
No. per Pack 100
Packaging Type Size K tubes with cap
Quantity 100 x 5 mL
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.