Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Lowenstein-Jensen Media

Catalog No. BB20909 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Packaging Type:
Slants (A tubes)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BB20909 100/Pk. Slants (A tubes)
BB20908 10/Pk. Slants (A tubes)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BB20909 Supplier BD Supplier No. 220909
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for cultivation of Mycobacterium tuberculosis and other mycobacterial species

  • Inspissated, egg-based media
  • Available as Mycoflask™ bottles, prepared slants and bottles

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2, EP, MCM9, SMWW
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Lowenstein-Jensen Media
Formulation Monopotassium Phosphate 2.5g/L, Magnesium Sulfate 0.24g/L, Sodium Citrate 0.6g/L, L-Asparagine 3.6g/L, Potato Flour 30g/L, Malachitegreen 0.4g/L, Glycerol 12mL, Whole Egg 1000mL
For Use With (Application) Cultivation of Mycobacterium tuberculosis and other mycobacterial species
Packaging Type Slants (A tubes)
Quantity 100/Pk.

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.