Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood

Catalog No. B21261X Shop All Waters Corporation Products
Change view
Click to view available options
Color:
Dark Red
Red
Media Type:
TSA II with 5% Sheep Blood
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Media Type
B21261X Dark Red TSA II with 5% Sheep Blood
B21239X Red TSA II with 5% Sheep Blood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B21261X Supplier BD Supplier No. 221261
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for cultivating fastidious microorganisms and for visualization of hemolytic reactions produced by many bacterial species

  • Ready for immediate use
  • Available with 5% and 10% sheep blood
  • “Deep-fill labelled plates contain additional volume of medium to reduce effects of drying during prolonged incubation”

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Red
Content And Storage 2°C to 8°C
Format Plate
Manufacturing Origin United States
Media Type TSA II with 5% Sheep Blood
Product Type Prepared Microbiology Plate
Target Organism Class Bacteria
Test Type Microbial Cultivation
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5% or 10%
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Incubation Conditions 35 ±2°C
Incubation Time 18 to 24 hr.
Packaging Type Plate
Quantity 100/Pk.
Height (Metric) 15 mm
Protein Source Casein, Soy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.