Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs for Veterinary Laboratory Use
Catalog No. L4391255 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Concentration:
30 μg
5 μg
Antimicrobial Agents:
Ceftiofur
Enrofloxacin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Antimicrobial Agents
L4391255 30 μg Ceftiofur
B4331717 5 μg Enrofloxacin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L4391255 Supplier BD Supplier No. 291255
Add to Cart
Add to Cart
This item is not returnable. View return policy
Same as standard Sensi-Disc Susceptibility Test Discs, but for veterinary laboratory use.
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Ceftiofur
Concentration 30 μg
Packaging Type Disc
Quantity 50 Discs