Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BBP™85 Series BradyGlo™ Phosphorescent Polyester Label Supply - Photoluminescent Green

Catalog No. 17111025 Shop All Brady Corporation Products
Change view
Click to view available options
BBP™85 Series BradyGlo™ Phosphorescent Polyester Label Supply - Photoluminescent Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-111-025 BBP™85 Series BradyGlo™ Phosphorescent Polyester Label Supply - Photoluminescent Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-111-025 Supplier Brady™ Supplier No. 105914
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BradyGlo™ B-526 High Intensity Tape is designed to provide high adhesion and luminance for egress marking in low light areas in and along stairwells of public buildings.

  • Ideal for exit or directional signs, fire alarms and extinguishers, egress pathway markings and directional systems for evacuation routes
  • Long lasting glow-in-the-dark identification glows up to 10 hours
  • Complies with RS 6-1 and 6-1A of New York Local Law 26 including photoluminescence, washability, flammability, toxic gas generation and radioactivity
  • Approved Model Numbers Accepted For Use, City of New York, Department of Buildings: MEA No. 467-06-M UV: 46-9-7

Specifications

Autoclavable Not Autoclavable
Description Label
Adhesive Type Permanent Acrylic
Color Glow
Finish Gloss
Format Roll
Material Phosphorescent Polyester
Print Technology Thermal Transfer
Product Type Label
For Use With (Equipment) BBP™85 industrial Sign and Label Printer, PowerMark™ industrial Sign and Label Printer
Quantity 1 Roll
Size (English) 33 ft. x 10 in.
Length (English) 33 ft.
Width (Metric) 130.81 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.