Learn More
Hanna Pharmaceutical Supply Co., Inc. IV EXTENSION SET 1.2 MICRON
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 473994
Encompass_Preferred
1.2 Micron Air-eliminating Filter with Luer Lock Connector. DEHP-free, Latex-free. Priming Volume: 4.3 mL, Length: 10 in. (25.4 cm) 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC1431455
 Restricted Item