Learn More
Hanna Pharmaceutical Supply Co., Inc. IV PUMP SET INFUSOMAT 136""
Catalog No. NC1660116
Encompass_Preferred
Infusomat Space Pump IV Set(60 drops/mL) Low adsorption, universal spike, free-flow protection clamp, roller clamp, SPIN-LOCK connector. Components are not made with DEHP or natural rubber latex. Priming volume: 24 mL, Length: 130 in. (330 cm) 24/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1660116 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BBR490037
Infusomat Space Pump IV Set(60 drops/mL) Low adsorption, universal spike, free-flow protection clamp, roller clamp, SPIN-LOCK connector. Components are not made with DEHP or natural rubber latex. Priming volume: 24 mL, Length: 130 in. (330 cm) 24/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.