Learn More
Bioworld BBS-Casein (10X), 500 mL
Catalog No. 50198540
Encompass_Preferred
BSS-Casein, supplied in 10X PBS and 5% Csein. Technical Information:10X Concentrate has a working pH of 8.5 ( +/- 0.1)Formulated in a borate buffered physiologic saline bufferSold as 2 x 100mL or 500mL.
Catalog No. 50-198-540 Supplier Bioworld Supplier No. 209600002
BSS-Casein, supplied in 10X PBS and 5% Csein. Technical Information:10X Concentrate has a working pH of 8.5 ( +/- 0.1)Formulated in a borate buffered physiologic saline bufferSold as 2 x 100mL or 500mL.