Learn More
Bioworld BBS/Fish Gelatin (10X), 200 mL
Catalog No. 50198396
Encompass_Preferred
BBS/Fish Gelatin 10XBBS Buffered fish gelatin 5% blocks (10X), diluents and coating solutions are supplied as 10X concentrates. Technical Information: pH range: 8.0 (+/- 0.1) formulated in a borate buffered physiologic saline bufferSold as 2 x 100mL or 500mL.
Catalog No. 50-198-396 Supplier Bioworld Supplier No. 207200001
BBS/Fish Gelatin 10XBBS Buffered fish gelatin 5% blocks (10X), diluents and coating solutions are supplied as 10X concentrates. Technical Information: pH range: 8.0 (+/- 0.1) formulated in a borate buffered physiologic saline bufferSold as 2 x 100mL or 500mL.