Learn More
Bioworld BBS/Glycerol/Casein, 500 mL
Supplier:  Bioworld 207200012
Encompass_Preferred
BBS/Glycerol/Casein Technical Information:10X Concentrate has a working pH of 7.8 (+/-) at 1:1010X Concentrate has a working pH of 8.2 (+/-) at 1:20Formulated in a borate buffered physiological saline buffer with glycerol as a preservative
Catalog No. 50-198-399