Learn More
Bioworld BetterBlocker™ Fish Gelatin in Glycerol/BBS (10X), 500 mL
Catalog No. 50198401
Encompass_Preferred
10 X Fish Gelatin BetterBlocker ™ with BBS and GlycerolBuffered fish gelatin 0.05% blocks, diluents and coating solutions are supplied as 10X concentrates.Sold as 2 x 100mL or 500mL.Technical Information:pH range: 8.0 (+/- 0.1) formulated in borate buffered physiologic saline buffer containing glycerol as a preservative
Catalog No. 50-198-401 Supplier Bioworld Supplier No. 207200022
10 X Fish Gelatin BetterBlocker ™ with BBS and GlycerolBuffered fish gelatin 0.05% blocks, diluents and coating solutions are supplied as 10X concentrates.Sold as 2 x 100mL or 500mL.Technical Information:pH range: 8.0 (+/- 0.1) formulated in borate buffered physiologic saline buffer containing glycerol as a preservative