Learn More
Aviva Systems Biology Corp BBS2 Antibody - middle region (ARP59751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BBS2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human BBS2. Alternate names: BBS, MGC20703ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-809