Learn More
Aviva Systems Biology Corp BBS4 Antibody - N-terminal region (ARP57695_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57695_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BBS4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BBS4. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-351