Learn More
Aviva Systems Biology Corp BBS5 Antibody - N-terminal region (ARP52930_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52930_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BBS5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BBS5. Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-788