Learn More
New England Biolabs, Inc. BbsI – 1500 units
SDP
Catalog No. 50812208
Encompass
A restriction endonuclease that recognizes the sequence GAAGACNN^NNNN_.
Catalog No. 50-812-208 Supplier New England Biolabs, Inc. Supplier No. R0539L
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A restriction endonuclease that recognizes the sequence GAAGACNN^NNNN_.