Learn More
New England Biolabs, Inc. BbsI – 300 units
SDP
Catalog No. 50812209
Encompass
A restriction endonuclease that recognizes the sequence GAAGACNN^NNNN_.
Catalog No. 50-812-209 Supplier New England Biolabs, Inc. Supplier No. R0539S
A restriction endonuclease that recognizes the sequence GAAGACNN^NNNN_.