Learn More
New England Biolabs, Inc. BbvCI – 100 units
SDP
Catalog No. 50812324
Encompass
A restriction endonuclease that recognizes the sequence CC^TCA_GC.
Catalog No. 50-812-324 Supplier New England Biolabs, Inc. Supplier No. R0601S
A restriction endonuclease that recognizes the sequence CC^TCA_GC.