Learn More
New England Biolabs, Inc. BbvCI – 500 units
SDP
Catalog No. 50812323
Encompass
A restriction endonuclease that recognizes the sequence CC^TCA_GC.
Catalog No. 50-812-323 Supplier New England Biolabs, Inc. Supplier No. R0601L
A restriction endonuclease that recognizes the sequence CC^TCA_GC.