Learn More
New England Biolabs, Inc. BbvI – 300 units
SDP
Catalog No. 50812085
Encompass
A restriction endonuclease that recognizes the sequence GCAGCNNNNNNNN^NNNN_.
Catalog No. 50-812-085 Supplier New England Biolabs, Inc. Supplier No. R0173S
A restriction endonuclease that recognizes the sequence GCAGCNNNNNNNN^NNNN_.