Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ BBXG (With Glycerol)

Catalog No. MP112333304 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP112333304 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112333304 Supplier MP Biomedicals™ Supplier No. 112333304

missing translation for 'validMainframeMsg'

Used as a loading dye for DNA samples. MP Biomedicals BBXG (With Glycerol) is a 10x solution of Bromophenol Blue, Xylene Cyanol FF and Glycerol that is used as a loading buffer for DNA gel electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Quantity 15 mL
Product Type DNA Loading Dye
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.