Learn More
Medchemexpress LLC BC-11 hydrobromide | 443776-49-6 | 99.0% | 290.97 | 50 MG
SDP
Catalog No. 5000347184
Encompass_Preferred
BC-11 hydrobromide is a selective TMPRSS2 inhibitor, which is a crucial host cellular factor for viral entry and SARS-CoV-2 pathogenesis. It also acts as a selective urokinase (uPA) inhibitor with an IC50 of 8.2 μM. This compound exhibits cytotoxicity towards triple-negative MDA-MB231 breast cancer cells, making it relevant for research on viral infections and cancer.
  • A selective TMPRSS2 inhibitor.
  • A selective urokinase inhibitor.
  • Cytotoxic to triple-negative MDA-MB231 breast cancer cells.
  • Used in research on viral infections and cancer.
Catalog No. 50-003-47184 Supplier Medchemexpress LLC Supplier No. HY10844710MG
BC-11 hydrobromide is a selective TMPRSS2 inhibitor, which is a crucial host cellular factor for viral entry and SARS-CoV-2 pathogenesis. It also acts as a selective urokinase (uPA) inhibitor with an IC50 of 8.2 μM. This compound exhibits cytotoxicity towards triple-negative MDA-MB231 breast cancer cells, making it relevant for research on viral infections and cancer.
  • A selective TMPRSS2 inhibitor.
  • A selective urokinase inhibitor.
  • Cytotoxic to triple-negative MDA-MB231 breast cancer cells.
  • Used in research on viral infections and cancer.