Learn More
Medchemexpress LLC BC-11 hydrobromide | 443776-49-6 | 99.0% | 290.97 g/mol | C8H12BBrN2O2S | 5 MG
SDP
Catalog No. 5000264763
Encompass_Preferred
BC-11 hydrobromide is a research-grade small-molecule inhibitor that targets TMPRSS2 and urokinase (uPA). It is used in biochemical and cellular studies of viral entry and cancer biology, and is supplied as a hydrobromide salt to aid stability and handling.
  • Selective TMPRSS2 and urokinase (uPA) inhibition for mechanistic studies.
  • Suitable for biochemical and cellular assays, including viral entry research.
  • Hydrobromide salt form for improved stability and solubility.
  • High purity (99.0%) and available in small research sizes (5-100 mg).
Catalog No. 50-002-64763 Supplier Medchemexpress LLC Supplier No. HY1084475MG
BC-11 hydrobromide is a research-grade small-molecule inhibitor that targets TMPRSS2 and urokinase (uPA). It is used in biochemical and cellular studies of viral entry and cancer biology, and is supplied as a hydrobromide salt to aid stability and handling.
  • Selective TMPRSS2 and urokinase (uPA) inhibition for mechanistic studies.
  • Suitable for biochemical and cellular assays, including viral entry research.
  • Hydrobromide salt form for improved stability and solubility.
  • High purity (99.0%) and available in small research sizes (5-100 mg).