Learn More
Abcam BC 11 hydrobromide, Urokinase (uPA) inhibitor, 10MG
Catalog No. 502706815
Encompass_Preferred
MW 290.97 g/mol, Purity >99%. Selective urokinase (uPA) inhibitor (IC₅₀ = 8.2 μM). Shows selectivity over 8 other related enzymes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6815 Supplier Abcam Supplier No. AB14119410MG
MW 290.97 g/mol, Purity >99%. Selective urokinase (uPA) inhibitor (IC₅₀ = 8.2 μM). Shows selectivity over 8 other related enzymes.
The product is subject to the following: Abcam Restricted Use Statement