Learn More
Abcam BC 11 hydrobromide, Urokinase (uPA) inhibitor, 50MG
Catalog No. 502706814
Encompass_Preferred
MW 290.97 g/mol, Purity >99%. Selective urokinase (uPA) inhibitor (IC₅₀ = 8.2 μM). Shows selectivity over 8 other related enzymes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6814 Supplier Abcam Supplier No. AB14119450MG
MW 290.97 g/mol, Purity >99%. Selective urokinase (uPA) inhibitor (IC₅₀ = 8.2 μM). Shows selectivity over 8 other related enzymes.
The product is subject to the following: Abcam Restricted Use Statement