Learn More
Cayman Chemical BC-11hydrobromIde 10mg
SDP
Catalog No. 502919192
Encompass_Preferred
A uPA inhibitor; reduces cell viability of MDA-MB-231 cells (EC50 = 117 µM)
Catalog No. 50-291-9192 Supplier Cayman Chemical Supplier No. 3952210 MG
A uPA inhibitor; reduces cell viability of MDA-MB-231 cells (EC50 = 117 µM)