Learn More
Selleck Chemical LLC BC-1215 S0322-25MG
SDP
Catalog No. 5000042213
Encompass_Preferred
BC-1215 is an inhibitor of F-box protein 3 (Fbxo3 a ubiquitin E3 ligase component) with IC50 of 0 9 g/mL and LC50 of 87 g/ml for IL-1 release BC-1215 inhibits the Fbxo3-TRAF activation pathway by destabilizing TRAF1-6
Catalog No. 50-000-42213 Supplier Selleck Chemical LLC Supplier No. S032225MG
BC-1215 is an inhibitor of F-box protein 3 (Fbxo3 a ubiquitin E3 ligase component) with IC50 of 0 9 g/mL and LC50 of 87 g/ml for IL-1 release BC-1215 inhibits the Fbxo3-TRAF activation pathway by destabilizing TRAF1-6